missing translation for 'onlineSavingsMsg'
Learn More

SLAMF2 Antibody, ProSci

Numéro de catalogue. 16594644
Modifier l'affichage
Click to view available options
Quantité:
20 μg Trial Size
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
16594644 20 μg Trial Size
16917457 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 16594644 Fournisseur ProSci Llc Code fournisseur 62490020

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Rabbit Polyclonal Antibody

Spécifications

Antigène SLAMF2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugué Unconjugated
Dilution SLAMF2 antibody can be used for detection of SLAMF2 by Western blot at 1 - 2 μg/mL. Antibody validated: Western Blot in rat samples. All other applications and species not yet tested.
Formule SLAMF2 Antibody is supplied in PBS containing 0.02% sodium azide.
Numéro d’acquisition du gène NP_1769
Alias du gène SLAMF2 Antibody: BCM1, BLAST, hCD48, mCD48, BLAST1, SLAMF2, MEM-102, BCM1, CD48 antigen, B-lymphocyte activation marker BLAST-1
Espèces hôtes Rabbit
Méthode de purification SLAMF2 Antibody is affinity chromatography purified via peptide column.
Quantité 20 μg Trial Size
Statut réglementaire RUO
Primaire ou secondaire Primary
ID du gène (Entrez) 962
Spécifications de test Two isoforms of SLAMF2 are known to exist; this antibody will recognize both isoforms. SLAMF2 antibody is predicted to not cross-react with other SLAM protein family members.
Espèces cibles Human, Mouse, Rat
Contenu et stockage SLAMF2 antibody can be stored at 4°C for three months and -20°C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
Type de produit Antibody
Forme Liquid
Isotype IgG
Catégorie de recherche Immunology
Afficher plus de résultats Afficher moins de résultats