missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals SKF-96365 hydrochloride, 99+%

Catalog No. AAJ60937MA
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60937MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ60937MA Supplier Thermo Scientific Chemicals Supplier No. J60937MA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Inhibits receptor-mediated calcium influx

Inhibits receptor-mediated calcium influx

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 130495-35-1
Molecular Formula C22H26N2O3·HCl
Synonym 1-[2-(4-Methoxyphenyl)-2-[3-(4-methoxyphenyl)propoxy]ethyl]imidazole

Specifications

Quantity 10 mg
Description White
For Use With (Application) Inhibits receptor-mediated calcium influx
Formula Weight 402.92g/mol
Solubility Soluble to 100mM in water
Product Type Signal Transduction Reagent
Form Powder
Assay ≥99%

RUO – Research Use Only