missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Single Stranded RNA (ssRNA) Fragments of SARS-CoV-2

Catalog No. EURM0191EA
Change view
Click to view available options
Content And Storage:
-20°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Content And Storage
EURM0191EA -20°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EURM0191EA Supplier MilliporeSigma™ Supelco™ Supplier No. EURM0191EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use as a positive control sample in testing for coronavirus

  • Used to verify correctness of transcription and amplification steps for SARS-CoV-2 real-time RT-PCR assays
  • Contains stabilized in vitro transcribed synthetic single stranded RNA
    • Does not contain any viable virus
  • Contains target regions that can be amplified by the following RT-PCR assays:
    • N1, N2 and N3 gene developed by Centers for Disease Control and Prevention (USA)
    • E gene and RdRP gene developed by Charité (DE)
    • N gene developed by Japanese Nationtional Institute of Infectious Disease (JP)
    • N gene developed by Ministry of Health of Thailand (TH)
    • S gene developed by Joint Research Centre of the European Commission (EU)

Specifications

Content And Storage -20°C
Description EURM reference material
Type Pharmacopeia & Metrological Institutes Standards
For Use With (Application) For use as a positive control sample in testing for the presence of the coronavirus