Learn More
Micro Essential Lab Single-Roll Hydrion™ pH Test Paper, With Chart
For measuring pH solutions.
Supplier:  Micro Essential Lab MES355
 View more versions of this product
Catalog No. 14853150H
Only null left
Description
Description
  • Used in laboratories, hospitals and universities worldwide
  • Comes with a 7/32 in. x 15 ft. roll of test paper and matching color chart
  • Provides for approximately 100 tests
Specifications
Chemical Identifiers
Colorimetric chart
100 Tests
pH Test Paper
Specifications
7.2 to 8.8
Colorimetric chart
pH Test Paper
For laboratories, hospitals and universities worldwide
100 Tests
Product Suggestions
Product Suggestions