Learn More
BRAND™ Single-Channel Micropipette Transferpette™ pro, Adjustable
Ergonomic single-channel micropipette with adjustable finger rest, easy calibration, and durable, autoclavable design.
$1009.02 - $1009.02
Specifications
Autoclavable Autoclavable
Disposable Reusable
Grip Type Ergonomic
Material Polypropylene
Number of Channels 1
View More Specs
Products 13
Catalog Number Mfr. No. Volume (Metric) Color Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Color Price Quantity  
13-200-170
View Documents
BRAND™
706868
1 μL Light Gray
Each for $1,009.02
Only null left
13-200-165
View Documents
BRAND™
706869
2.5 μL Light Gray
Each for $1,009.02
Only null left
13-200-186
View Documents
BRAND™
706870
10 μL Light Gray
Each for $1,009.02
Only null left
13-200-164
View Documents
BRAND™
706871
20 μL Light Gray
Each for $1,009.02
Only null left
13-200-202
View Documents
BRAND™
706872
20 μL Yellow
Each for $1,009.02
Only null left
13-200-181
View Documents
BRAND™
706873
50 μL Yellow
Each for $1,009.02
Only null left
13-200-182
View Documents
BRAND™
706874
100 μL Yellow
Each for $1,009.02
Only null left
13-200-206
View Documents
BRAND™
706878
200 μL Yellow
Each for $1,009.02
Only null left
13-200-167
View Documents
BRAND™
706879
300 μL Green
Each for $1,009.02
Only null left
13-200-180
missing translation for 'viewDocuments'
BRAND™
706880
1000 μL Dark Blue
Each for $1,009.02
Only null left
13-200-191
missing translation for 'viewDocuments'
BRAND™
706881
2500 μL Red
Each for $1,009.02
Only null left
13-200-162
missing translation for 'viewDocuments'
BRAND™
706882
5000 μL Red
Each for $1,009.02
Only null left
13-200-197
missing translation for 'viewDocuments'
BRAND™
706884
10,000 μL Blue
Each for $1,009.02
Only null left
Description
Description
  • Its slim design and adjustable finger rest ensure that the pipette fits perfectly in your hand, making it ergonomic and comfortable to use, even during long pipetting sessions
  • To keep you flexible at all times, the Transferpette™ pro features two adjustment techniques:
    • Easy calibration for permanent calibration without tools
    • User adjustment for quick, reversible adaptation to changing conditions such as temperature, tip shape, or media properties
  • The Transferpette™ pro is made of high-quality materials with built-in counter protection, making it both robust and durable. It's fully autoclavable (121°C) and available as single- and multi-channel pipettes.
Limitations of use:
  • Operating temperature should be between +15°C and +40°C (59°F to 104°F). Other temperatures upon request.
  • Vapor pressure up to 500 mbar
  • Viscosity: 260 mPa·s
  • For viscous media, the speed must be adjusted if necessary
Specifications
Specifications
Autoclavable
Ergonomic
1
Non-sterile
Performance Certificate, Shelf Mount, and Silicone Grease
Reusable
Polypropylene
No
Mechanical Pipette