missing translation for 'onlineSavingsMsg'
Learn More

Simport™ Scientific Tamper Evident Self-Standing Transport Tubes

Catalog No. 22040248
Change view
Click to view available options
Capacity (Metric):
5 mL
7 mL
10 mL
12 mL
30 mL
Graduated:
No
Yes
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Graduated
22040248 30 mL Yes
22040237 5 mL Yes
22040239 10 mL Yes
22040240 12 mL Yes
22040247 7 mL No
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 22040248 Supplier Simport™ Scientific Supplier No. T55230ATTP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Tamper Evident Self-Standing Transport Tubes for all applications requiring tamper evident seal for utmost security and sample integrity.

  • For secure short and long term storage
  • Provide strength and clarity
  • Non-toxic, non sterile polypropylene made etched or non graduated tubes
  • Exhibit unique design features
  • Unique transport tubes incorporate a seal that needs to be broken in order to open it, which leaves visual indication that the vial has been opened
  • With external threads for providing smooth and uniform inner surface
  • Closure offers unique frangible band design
  • Combination of tamper evident and resistant closure system
  • For clinical trials and expensive reagents in diagnostic kits
  • Caps sold separately

Specifications

Autoclavable Autoclavable
Capacity (Metric) 30 mL
Bottom Shape Conical
Material Polypropylene
For Use With (Application) Storage and transportation of biological materials
Length (Metric) 111 mm
Sterility Non-sterile
Graduated Yes
Packaging Quantity 500/Cs.
Closure Material Polyethylene
Capacity (English) 1.01 oz.
Diameter (Metric) 25.3 mm
Product Type Transport Tube (Self-Standing)
Show More Show Less