missing translation for 'onlineSavingsMsg'
Learn More

Silvex™ 2-(2,4,5-Trichlorophenoxy)propionic acid, MilliporeSigma™ Supelco™

Catalog No. 111016708
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016708 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111016708 Supplier Merck Emd Millipore Supplier No. 45691100MG
Agriculture

Specifications

Product Line Silvex
CAS 93-72-1
Physical Form Neat
Linear Formula C9H7 Cl3O3
MDL Number MFCD00002646
Synonym 2,4,5-TP; Fenoprop; Silvex™
Type Analytical Standard
Quantity 100 mg
Formula Weight 269.51
Grade Analytical Standard

Silvex is a registered trademark of Silberline Manufacturing Co., Inc.