Learn More
Alkali Scientific SilverSeal II™ Aluminum Sealing Film, 100/pk
Numéro de catalogue. 502388901
Encompass
Compared to other aluminum foils, Aluminum Foil Sealing Film has less tendency to roll back on itself when removing the backing paper. Certified DNase- and RNase- free. Excellent chemical resistance (e.g. DMSO). Excellent chemical resistance (e.g. DMSO). 1.4mil conformable aluminum foil allows visual verification of the seals security around each well. End-tab removal before thermal cycling is highly recommended. End-tab removal before thermal cycling is highly recommended.
Numéro de catalogue. 502388901 Fournisseur Alkali Scientific Code fournisseur SF2SS
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Compared to other aluminum foils, Aluminum Foil Sealing Film has less tendency to roll back on itself when removing the backing paper. Certified DNase- and RNase- free. Excellent chemical resistance (e.g. DMSO). Excellent chemical resistance (e.g. DMSO). 1.4mil conformable aluminum foil allows visual verification of the seals security around each well. End-tab removal before thermal cycling is highly recommended. End-tab removal before thermal cycling is highly recommended.