missing translation for 'onlineSavingsMsg'
Learn More

Silver Wool, For Elementary Analysis, MilliporeSigma™

Catalog No. M1015060010
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1015060010 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1015060010 Supplier Technidata Supplier No. 1.01506.0010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
PubChem CID 23954
ChEBI CHEBI:9141
IUPAC Name silver
SMILES [Ag]

Specifications

Appearance Silver colored solid
Quantity 10 g
Odor Odorless
Packaging Glass bottle
Form Solid
Formula Weight 107.86
Chemical Name or Material Silver Wool