missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfate d’argent, grade ACS, ≥ 98,0%, LabChem™
Spécifications
Spécifications
| Nom ou substance chimique | Silver Sulfate |
| Point de fusion | 652 °C |
| Couleur | Blanc |
| CAS % maximal | 100 |
| Forme physique | Cristalline |
| Quantité | 25 g |
| Formule linéaire | Ag2SO4 |
| Renseignements sur la solubilité | Soluble dans l’eau |
| Poids de formule | 311.79 |
| Pourcentage de pureté | ≥98.0% |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye damage. Very toxic to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement Wear eye protection, protective gloves, face protection. Wash exposed skin thoroughly after handling. Avoid release to the environment. If on skin: Wash with plenty of soap and water. Take off contaminated clothing and wash before reuse. If skin irritation occurs: Get medical advice/attention. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a poison center/doctor. Collect spillage. Dispose of contents/container to comply with local, state and federal regulations. |
| EINECSNumber | 233-653-7 |
| Recommended Storage | Température ambiante |