missing translation for 'onlineSavingsMsg'
Learn More

Silver shot, 1-7mm (0.04-0.1in), Premion™, 99.999% (metals basis)

Catalog No. AA1135730
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1135730 250 g
AA1135709 10 g
AA1135718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1135730 Supplier Thermo Scientific Chemicals Supplier No. 01135730

For vapor deposition | CAS: 7440-22-4 | Ag | 107.87 g/mol

For vapor deposition

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
PubChem CID 23954
ChEBI CHEBI:9141
IUPAC Name silver
SMILES [Ag]

Specifications

Quantity 250 g
Odor Characteristic
Form Shot
Percent Purity 99.999%
Grade Premion
Assay (metals basis)
Chemical Name or Material Silver shot, 1 to 3mm (0.04 to 0.1 in.)
EINECSNumber 231-131-3
RTECSNumber VW3500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only