missing translation for 'onlineSavingsMsg'
Learn More

Silver perchlorate monohydrate, 99.9% (metals basis)

Catalog No. AA1162404
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1162404 2 g
AA1162409 10 g
AA1162418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1162404 Supplier Thermo Scientific Chemicals Supplier No. 01162404

CAS: 14242-05-8 | AgClO4 | 207.31 g/mol

Silver perchlorate can be used in the manufacturing of explosives. It is generally used as a source of silver ions during silver nanoparticles synthesis. It is used as a catalyst in organic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14242-05-8
Molecular Formula AgClO4
Molecular Weight (g/mol) 207.31
MDL Number MFCD00149128
InChI Key YDHABVNRCBNRNZ-UHFFFAOYSA-M
Synonym silver perchlorate hydrate, silver perchlorate monohydrate, silver perchlorite, silver perchloric acid, acmc-1brvv, ag.clo4.h2o, silver i perchlorate monohydrate, silver 1+ ion hydrate perchlorate, silver 1+ hydrate perchlorate
PubChem CID 15959344
IUPAC Name silver;perchlorate;hydrate
SMILES [Ag+].[O-][Cl](=O)(=O)=O

Specifications

Melting Point 485°C (decomposition)
Color White
Physical Form Crystalline
Quantity 2 g
UN Number UN1481
Merck Index 14,8523
Solubility Information Soluble in water and organic solvents.
Formula Weight 225.34 (207.32 Anhydrous)
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic; Light sensitive
Density 2.800 g/mL
Assay (metals basis)
Chemical Name or Material Silver perchlorate monohydrate
Show More Show Less
Hazard Category H272-H314
Hazard Statement GHS H Statement
H272-H314-H318
May intensify fire
oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P220-P221-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.
EINECSNumber 232-035-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only