missing translation for 'onlineSavingsMsg'
Learn More

Silver perchlorate, anhydrous

Catalog No. AA4220922
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4220922 100 g
AA4220906 5 g
AA4220914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4220922 Supplier Thermo Scientific Chemicals Supplier No. 04220922

Light Sensitive & Hygroscopic | CAS: 7783-93-9 | AgClO4 | 207.314 g/mol

Silver perchlorate is used as a catalyst in organic chemistry. It is an effective reagent for replacing halides ligands with perchlorate. It is also used in the manufacturing of explosives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7783-93-9
Molecular Formula AgClO4
Molecular Weight (g/mol) 207.314
MDL Number MFCD00003400
InChI Key YDHABVNRCBNRNZ-UHFFFAOYSA-M
Synonym silver perchlorate, unii-sv4x1u113o, silver 1+ ion perchlorate, perchloric acid, silver 1+ salt, silver 1+ salt, ag.clo4, perchloric acid, silver 1+ salt 1:1, silver 1+ perchlorate ion, silver i perchloratehydrate, silver i perchlorate, anhydrous
PubChem CID 24562
IUPAC Name silver;perchlorate
SMILES [O-]Cl(=O)(=O)=O.[Ag+]

Specifications

Melting Point 485°C (decomposition)
Color White
Physical Form Crystals
Quantity 100 g
Solubility Information Soluble in water and many organic solvents (benzene,toluene,pyridine)Soluble in water,benzene and toluene.
Formula Weight 207.32
Sensitivity Hygroscopic; Light sensitive
Appearance White crystals
Density 2.806
Chemical Name or Material Silver perchlorate
Hazard Category H272-H314-H335
Precautionary Statement P210-P220-P221-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation DOT Class: 5.1, Packaging Group: II
EINECSNumber 232-035-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only