Learn More
Chemical Identifiers
| CAS | 7761-88-8 |
|---|---|
| Molecular Formula | AgNO3 |
| Molecular Weight (g/mol) | 169.87 |
| MDL Number | MFCD00003414 |
| InChI Key | SQGYOTSLMSWVJD-UHFFFAOYSA-N |
| Synonym | silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol |
| PubChem CID | 24470 |
| ChEBI | CHEBI:32130 |
| IUPAC Name | silver(1+) nitrate |
| SMILES | [Ag+].[O-][N+]([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
SS816-4
|
Thermo Fisher Scientific
SS8164 |
4 L | Glass Bottle |
Each for $5,532.75
Case of 4 Each for $22,131.00
|
|
||||
Description
EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.
Chemical Identifiers
| 7761-88-8 | |
| 169.87 | |
| SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
| 24470 | |
| silver(1+) nitrate |
| AgNO3 | |
| MFCD00003414 | |
| silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
| CHEBI:32130 | |
| [Ag+].[O-][N+]([O-])=O |
Specifications
| Colorless | |
| AgNO3 | |
| 0.245 to 0.255N | |
| SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
| silver(1+) nitrate | |
| 24470 | |
| 169.87 | |
| Glass Bottle | |
| 14mmHg at 20°C | |
| 4 L | |
| Silver Nitrate Solution, 0.25N |
Safety and Handling
WARNING!
Emergency Overview
Irritating to eyes and skin. Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Wear personal protective equipment. Ensure adequate ventilation. Avoid contact with skin, eyes and clothing. Avoid ingestion and inhalation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur.
NFPA
Health:2
Flammability:0
Instability:0
DOTInformation : DOT Class 5.1, : Oxidizer