missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate Solution (0.1N (N/10)/Certified), Fisher Chemical™ Edge
Edge

Standardized at 25°C against NIST™ potassium chloride

Supplier:  Thermo Fisher Scientific FLSS721

 View more versions of this product

Catalog No. SS72-1



Only null left
Edge
Explore available promotions

Description

Description

Specifications

Specifications

Silver Nitrate Solution
0.1N (N/10)
AgNO3
3082
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
[Ag+].[O-][N+]([O-])=O
169.87
CHEBI:32130
Pass Test
999b POTASSIUM CHLORIDE
1 L
Colorless
7761-88-8 , 7732-18-5
MFCD00003414
0.0995 to 0.1005N
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver(1+) nitrate
24470
Certified
Glass Bottle
14mmHg at 20°C
Liquid

Chemical Identifiers

7761-88-8
169.87
SQGYOTSLMSWVJD-UHFFFAOYSA-N
24470
silver(1+) nitrate
AgNO3
MFCD00003414
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
CHEBI:32130
[Ag+].[O-][N+]([O-])=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
May cause eye, skin, and respiratory tract irritation . The toxicological properties have not been fully investigated. Harmful to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
NFPA
Health:1 Flammability:0 Instability:0

Signal Word: CAUTION!

DOTInformation : DOT Class 5.1, : Oxidizer

SDS Documents

Documents

Promotions

Promotions