missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate, in Isopropanol, Certified, 0.0100N ±0.0002N (0.01M), LabChem™

Catalog No. p-5388215
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC227451 500 mL
LC227452 1 L
LC227454 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC227451 Supplier LabChem Supplier No. LC227451
Only null left

Chemical Identifiers

CAS 7761-88-8 , 67-63-0 , 7732-18-5
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Boiling Point 83°C
Melting Point -88°C
Color Colorless
CAS Max % 0.17, 99.73, 0.1
Physical Form Liquid
Quantity 500 mL
Linear Formula AgNO3
UN Number UN1993
Flash Point 12°C
Solubility Information Soluble in water
Formula Weight 169.87
Grade Certified
Concentration or Composition (by Analyte or Components) 0.0100N ±0.0002N (0.01M)
Identification Passes Test
Packaging Amber Glass
Specific Gravity 0.8g/cm3
Decomposition Information Fume; Carbon monoxide; Carbon dioxide; May release flammable gases
Density 0.8g/cm3
Chemical Name or Material Silver Nitrate, in Isopropanol
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Keep away from heat, hot surfaces, open flames, sparks.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, lighting, ventilating equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Avoid breathing mist, vapors, spray.
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
In case of fire: Use carbon dioxide (CO2), powder, alcohol-resistant foam to extinguish.
Store locked up a cool, well ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature