Learn More
Electron Microscopy Sciences Silver Nitrate 10% Aqueous 250 ML
Numéro de catalogue. 5031890
Encompass
Used in Hirano-Zimmerman Method for Nerve Cells and Fibers for staining of nerofibrils, dendrites, axis cylinders, senile plaques, etc. See AFIP Manual of Histological Staining Methods, 3rd ed., Ed. L.Luna: New York: McGraw-Hill Pub., c 1968, p. 198 or Hirano, A. and Zimmerman, H.M., Arch. Neurol., 6:114-122, 1962
Numéro de catalogue. 5031890 Fournisseur Electron Microscopy Sciences Code fournisseur 2669801
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Used in Hirano-Zimmerman Method for Nerve Cells and Fibers for staining of nerofibrils, dendrites, axis cylinders, senile plaques, etc. See AFIP Manual of Histological Staining Methods, 3rd ed., Ed. L.Luna: New York: McGraw-Hill Pub., c 1968, p. 198 or Hirano, A. and Zimmerman, H.M., Arch. Neurol., 6:114-122, 1962