missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Nitrate, 0.100 N (N/10), Ricca Chemical
Silver Nitrate Titrants
$133.50 - $4363.10
Chemical Identifiers
| CAS | 7761-88-8 |
|---|---|
| Molecular Formula | AgNO3 |
| Molecular Weight (g/mol) | 169.87 |
| MDL Number | MFCD00003414 |
| InChI Key | SQGYOTSLMSWVJD-UHFFFAOYSA-N |
| Synonym | silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol |
| PubChem CID | 24470 |
| ChEBI | CHEBI:32130 |
| IUPAC Name | silver(1+) nitrate |
| SMILES | [Ag+].[O-][N+]([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
7000-4
|
Ricca Chemical Company
RI70004 |
120 mL | Amber Poly Bottle |
Each for $133.50
|
|
||||
|
7000-16
|
Ricca Chemical Company
700016 |
500 mL | Amber Poly Bottle |
Each for $223.61
|
|
||||
|
R70000001C
|
Ricca Chemical Company
R70000001C |
1 L | Amber Glass Bottle |
Each for $469.83
|
|
||||
|
7000-32
|
Ricca Chemical Company
700032 |
1 L | Amber Poly Bottle |
Each for $347.80
|
|
||||
|
R70000004C
|
Ricca Chemical Company
R70000004C |
4 L | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
7000-1
|
Ricca Chemical Company
70001 |
4 L | Black Poly Bottle |
Each for $1,252.83
|
|
||||
|
R70000004F
|
Ricca Chemical Company
R70000004F |
4 L | Cubitainer |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
7000-5
|
Ricca Chemical Company
70005 |
20 L | Cubitainer |
Each for $4,363.10
|
|
||||
Chemical Identifiers
| 7761-88-8 | |
| 169.87 | |
| SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
| 24470 | |
| silver(1+) nitrate |
| AgNO3 | |
| MFCD00003414 | |
| silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
| CHEBI:32130 | |
| [Ag+].[O-][N+]([O-])=O |
Specifications
| 7761-88-8,7732-18-5 | |
| MFCD00003414 | |
| SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
| silver(1+) nitrate | |
| 24470 | |
| Laboratory | |
| Amber Poly Bottle | |
| 120 mL |
| AgNO3 | |
| silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
| [Ag+].[O-][N+]([O-])=O | |
| 169.87 | |
| CHEBI:32130 | |
| 0.100N | |
| ∼6 | |
| Silver Nitrate Solution |
Safety and Handling
ShelfLife : 24 months
Recommended Storage : Ambient