missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate, 0.100 N (N/10), Ricca Chemical

Silver Nitrate Titrants

Supplier:  Ricca Chemical Company 70001

 View more versions of this product

Catalog No. 7000-1


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Silver Nitrate Solution
AgNO3
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
[Ag+].[O-][N+]([O-])=O
169.87
CHEBI:32130
0.100N
∼6
7761-88-8,7732-18-5
MFCD00003414
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver(1+) nitrate
24470
Laboratory
Black Poly Bottle
4 L

Chemical Identifiers

7761-88-8
169.87
SQGYOTSLMSWVJD-UHFFFAOYSA-N
24470
silver(1+) nitrate
AgNO3
MFCD00003414
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
CHEBI:32130
[Ag+].[O-][N+]([O-])=O
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions