missing translation for 'onlineSavingsMsg'
Learn More

Silver iodide, Premion™, 99.999% (metals basis)

Catalog No. AA1211106
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1211106 5 g
AA1211114 25 g
AA1211122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1211106 Supplier Thermo Scientific Chemicals Supplier No. 01211106
Only null left

CAS: 7783-96-2 | AgI | 234.773 g/mol

Silver iodide is used in photographic film. Used in medical devices as a sustained release antibacterial. It also used in as an antiseptic and in cloud seeding. It is used in proteomics research applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7783-96-2
Molecular Formula AgI
Molecular Weight (g/mol) 234.773
MDL Number MFCD00003412
InChI Key MSFPLIAKTHOCQP-UHFFFAOYSA-M
Synonym silver iodide, neosiluol, neosilvol, silver monoiodide, silver 1+ iodide, colloidal silver iodide, iodanylsilver, silver iodide metals basis, ccris 8490
PubChem CID 24563
IUPAC Name iodosilver
SMILES [Ag]I

Specifications

Melting Point 558°C
Color Yellow
Physical Form Powder
Quantity 5 g
UN Number UN3077
Merck Index 14,8516
Solubility Information Soluble in alkali cyanides and iodides. Insoluble in water,acids and ammonium carbonate.
Formula Weight 234.77
Percent Purity 99.999%
Grade Premion
Odor Odorless
Sensitivity Light sensitive
Density 5.67 g/mL
Assay (metals basis)
Chemical Name or Material Silver iodide
Show More Show Less
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 232-038-0
RTECSNumber VW4450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only