Learn More
Silver(II) fluoride, 98+%
Description
Silver(II) fluoride is used as a fluorinating and oxidizing agent. It is also used in the preparation of organic perfluorocompounds and in the fluorination of aromatic compounds. In addition, it is also used in pharmacy and electroplating industry. It serves as an intermediate in the catalysis of gaseous reactions with fluorine by silver. It is utilized in the oxidation of xenon to xenon difluoride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 690°C |
| Color | Brown |
| Physical Form | Crystalline |
| Quantity | 50 g |
| Assay Percent Range | 98+% |
| Solubility Information | Soluble in hexane,chloroform,chlorofluorocarbon and hydrogen fluoride. |
| Formula Weight | 145.87 |
| Sensitivity | Light and moisture sensitive |
| Appearance | Brown crystals |
| Density | 4.6 |
| Show More |
Safety and Handling
| Hazard Category | H272-H302+H312+H332-H314-H335 |
| Precautionary Statement | P210-P220-P221-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | DOT Class: 8, Packaging Group: II |
| EINECSNumber | 232-037-5 |
| RTECSNumber | VW4200000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only