missing translation for 'onlineSavingsMsg'
Learn More

Silver diethyldithiocarbamate, ACS

Catalog No. AA1189818
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1189818 50 g
AA1189809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1189818 Supplier Thermo Scientific Chemicals Supplier No. 01189818

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1470-61-7 | C5H10AgNS2 | 256.13 g/mol

Silver diethyldithiocarbamate cluster plays a useful role for the deposition of acanthite (Ag2S) thin films which is employed in photoelectrochemical applications. Silver diethyldithiocarbamate membrane is used to develop ion selective electrodes for nitrate ion. It is useful in the colorimetric determination of arsenic. It acts as a copper chelator, and is utilized to quantitate copper in biological materials by ESR. It also works as an inhibitor of superoxide dismutase, ascorbate oxidase, and other enzymes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1470-61-7
Molecular Formula C5H10AgNS2
Molecular Weight (g/mol) 256.13
MDL Number MFCD00004929
InChI Key NSVHDIYWJVLAGH-UHFFFAOYSA-M
Synonym silver diethyldithiocarbamate, unii-gvn001b86o, agddtc, silver n,n-diethylcarbamodithioate, diethyldithiocarbamic acid, silver salt, diethyl dithio carbamic acid silver salt, silver i diethyldithiocarbamate, silver, diethylcarbamodithioato-ks,ks', ditiocarb silver, silver n,n-diethyldithiocarbamate
PubChem CID 3034078
IUPAC Name silver;N,N-diethylcarbamodithioate
SMILES CCN(CC)C(=S)[S-].[Ag+]

Specifications

Melting Point 172°C to 175°C
Color White
Physical Form Powder
Quantity 50 g
Solubility Information It is soluble in water and pyridine.
Formula Weight 256.14
Grade ACS
Odor Odorless
Sensitivity Light sensitive
Appearance Yellow to pale brown powder
Chemical Name or Material Silver diethyldithiocarbamate, For analysis ACS
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 216-003-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only