missing translation for 'onlineSavingsMsg'
Learn More

Silver, 99.9%, (trace metal basis), wire, 1.0 mm diam., 1ea = 8.2gr

Catalog No. AC317740010
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC317740010 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC317740010 Supplier Thermo Scientific Chemicals Supplier No. 317740010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-22-4 | Ag | 107.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis

Chemical Identifiers

CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
PubChem CID 23954
ChEBI CHEBI:9141
IUPAC Name silver
SMILES [Ag]

Specifications

Color Silver
Melting Point 961.0°C
Boiling Point 2212.0°C
Quantity 1 Ea.
Form Wire
Assay Percent Range 99.9%
Linear Formula Ag
Fieser 04,164
Merck Index 15, 8643
Formula Weight 107.87
Percent Purity 99.9%
Grade Trace Metal Basis
Chemical Name or Material Silver, 99.9% (1ea = 8.2gr)
Show More Show Less

RUO – Research Use Only