Learn More
Micronova™ SilSat™ Silicone Wipes
GREENER_CHOICE
SilSat™ Silicone Wipes are designed for use in aseptic processing and support areas to lubricate gaskets and seals in isolators, refrigerators, and cabinets.
Supplier:  Micronova™ SSW604IR
Catalog No. 19166386
Only null left
Description
Description
  • Sterile, pre-saturated wipes
  • Designed to lubricate gaskets, seals and equipment
  • Gamma irradiated for use in aseptic areas
  • Processed for cleanrooms and controlled environments, ISO 5 to ISO 9
  • Good compatibility with common disinfectants and cleaning solutions
Specifications
Specifications
Pre-saturated Wipe
15.2 cm
10 cm
Good compatibility with common disinfectants and cleaning solutions
ISO 5 to ISO 9
Store at ambient temperature. Avoid exposure to direct sunlight. 18 Months shelf life (inclusive of manufacturing date).
Aseptic processing and support areas equipment
50% Silicone, 50% Alcohol
0.5 in.
6 in.
4 in.
Polyester
Not Autoclavable
Resealable Packages in Poly Lined Cases
6 x 4 in.
4 Wipes/Pk., 48 Pk./Cs.
Sterile
1.2 cm
Product Suggestions
Product Suggestions