Clearco Products Co Silicone Grease 3005, 5.3oz tube
Numéro de catalogue. NC0866201
This item has a minimum order quantity of 12 tubes per supplier requirements. SILICONE GREASE 3005 NSF H1 FOOD GRADE Chemical Name: Polydimethylsiloxane Compound /PDMS Silicone Grease Compound; 63148-62-9 Stiffness Rating: NLGI#2
Numéro de catalogue.
NC0866201
Fournisseur
Clearco Products CoCode fournisseur
63148629
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
This item has a minimum order quantity of 12 tubes per supplier requirements. SILICONE GREASE 3005 NSF H1 FOOD GRADE Chemical Name: Polydimethylsiloxane Compound /PDMS Silicone Grease Compound; 63148-62-9 Stiffness Rating: NLGI#2