missing translation for 'onlineSavingsMsg'
Learn More

Silicon rod, 5.08cm (2.0 in.) dia., 99.999% (metals basis), Thermo Scientific™

Catalog No. AA39103DM
Change view
Click to view available options
Quantity:
25 mm
100 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA39103DM 100 mm
AA39103DH 25 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA39103DM Supplier Thermo Scientific Chemicals Supplier No. 39103DM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 7440-21-3
Molecular Formula Si
Molecular Weight (g/mol) 28.09
MDL Number MFCD00085311
InChI Key XUIMIQQOPSSXEZ-UHFFFAOYSA-N
Synonym metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831
PubChem CID 5461123
ChEBI CHEBI:27573
IUPAC Name silicon
SMILES [Si]

Specifications

Quantity 100 mm
Odor Odorless
Weight ≈47.18g/cm
Form Rod
Percent Purity 99.999%
Assay (metals basis)
Chemical Name or Material Silicon rod, 5.08cm (2.0 in.) dia.
EINECSNumber 231-130-8
TSCA Yes
Recommended Storage Ambient temperatures