missing translation for 'onlineSavingsMsg'
Learn More

Silicon powder, crystalline, -325 mesh, 99.999% (metals basis)

Catalog No. AA3566218
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3566218 50 g
AA3566209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3566218 Supplier Thermo Scientific Chemicals Supplier No. 03566218

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-21-3 | Si | 28.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-21-3
Molecular Formula Si
Molecular Weight (g/mol) 28.09
MDL Number MFCD00085311
InChI Key XUIMIQQOPSSXEZ-UHFFFAOYSA-N
Synonym metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831
PubChem CID 5461123
ChEBI CHEBI:27573
IUPAC Name silicon
SMILES [Si]

Specifications

Color White
Quantity 50 g
Mesh Size −325 Mesh
Odor Odorless
Form Crystalline Powder
UN Number UN1346
Percent Purity 99.999%
Assay (metals basis)
Chemical Name or Material Silicon powder
Hazard Category H228
Hazard Statement GHS H Statement
H228
Flammable solid.
Precautionary Statement P210-P240-P241-P280-P370+P378q
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: SILICON POWDER, AMORPHOUS
EINECSNumber 231-130-8
RTECSNumber VW0400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only