missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silicon(II) oxide, 99.9% (metals basis)
Description
For vacuum deposition
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 1880°C |
| Melting Point | >1702°C |
| Physical Form | Pieces |
| Quantity | 500 g |
| Packaging | Plastic bottle |
| Specific Gravity | 2.13 |
| Density | 2.13 |
| Formula Weight | 44.09 |
| Percent Purity | 99.9% |
| Chemical Name or Material | Silicon(II) oxide |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 233-232-8 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only