missing translation for 'onlineSavingsMsg'
Learn More

Silicon ICP Standard, 10,000 ppm Si in H2O/tr HF, Ricca Chemical

Supplier:  Ricca Chemical Company PSI10KW50

Catalog No. PSI10KW50

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Silicon ICP Standard
16919-19-0 , 7664-39-3 , 7732-18-5
92.0, 5.84, 0.09
1 mL = 10 mg Si
ITHIMUMYFVCXSL-UHFFFAOYSA-P
diazanium;hexafluorosilicon(2-)
28145
Natural Poly Bottle
10,000ppm Si
96.0, 6.08, 0.1
F6H8N2Si
[NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F
178.153
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents