missing translation for 'onlineSavingsMsg'
Learn More

Silicon AA Standard, Certified, 1000ppm Si ±10ppm (1mL = 1mg Si), LabChem™

Catalog No. LC224007
Change view
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC224007 125 mL Poly Bottle
LC224001 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC224007 Supplier LabChem Supplier No. LC224007
Only null left

Chemical Identifiers

CAS 13517-24-3 , 7732-18-5
Molecular Formula H18Na2O12Si
Molecular Weight (g/mol) 284.197
InChI Key PHIQPXBZDGYJOG-UHFFFAOYSA-N
PubChem CID 61639
IUPAC Name disodium;dioxido(oxo)silane;nonahydrate
SMILES O.O.O.O.O.O.O.O.O.[O-][Si](=O)[O-].[Na+].[Na+]

Specifications

Packaging Poly Bottle
CAS Max % 98.99, 1.01
Identification Passes Test
Density 1g/mL
Physical Form Liquid
Concentration 1000 ppm Si ±10 ppm (1mL = 1mg Si)
Linear Formula Na2SiO3·9H2O
Color Colorless
Solubility Soluble in water
Quantity 125 mL
Formula Weight 284.2
Grade Certified
Chemical Name or Material Silicon AA Standard, 1000ppm (1mL = 1mg Si)
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Recommended Storage Room Temperature