missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silicon, 100-200 Mesh, Powder, 98%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Physical Form | Crystalline Powder or Needles |
| Quantity | 250 g |
| Molecular Weight (g/mol) | 28.09 |
| Percent Purity | 98% |
| Grade | Not Available |
| Packaging | Poly Bottle |
| Chemical Name or Material | Silicon, 100-200 Mesh, Powder |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.