missing translation for 'onlineSavingsMsg'
Learn More

Silica TLC, 60Å, EcoChrom™, MP Biomedicals™

Catalog No. ICN04671
Change view
Click to view available options
Quantity:
25 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN04671 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN04671 Supplier MP Biomedicals Inc Supplier No. 0204671
Only null left

For Chromatography; Chemical Synthesis

MP EcoChrom™ Silica for TLC are produced from the same highly standardized raw material as the MP EcoChrom™ Silica for column chromatography. They are available with or without binder and with or without fluorescent indicators

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name silanedione
SMILES O=[Si]=O

Specifications

Color White to Gray
Physical Form Powder
Quantity 25 kg
Formula Weight 60.08
Density 2g/cm3 at 20°C
Chemical Name or Material Silica, TLC
Recommended Storage Store at Room Temperature.