missing translation for 'onlineSavingsMsg'
Learn More

Silica Standard, Certified, 1000ppm ±10ppm (1mL = 1mg SiO2), LabChem™

Catalog No. LC223907
Change view
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC223907 125 mL Poly Bottle
LC223901 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC223907 Supplier LabChem Supplier No. LC223907
Only null left

Chemical Identifiers

CAS 13517-24-3 , 7732-18-5
Molecular Formula H18Na2O12Si
Molecular Weight (g/mol) 284.197
InChI Key PHIQPXBZDGYJOG-UHFFFAOYSA-N
PubChem CID 61639
IUPAC Name disodium;dioxido(oxo)silane;nonahydrate
SMILES O.O.O.O.O.O.O.O.O.[O-][Si](=O)[O-].[Na+].[Na+]

Specifications

Packaging Poly Bottle
CAS Max % 99.53, 0.47
Identification Passes Test
Density 1g/mL
Physical Form Liquid
Concentration 1000 ppm ±10 ppm (1mL = 1mg SiO2)
Linear Formula Na2SiO3·9H2O
Color Colorless
Solubility Soluble in water
Quantity 125 mL
Formula Weight 284.2
Grade Certified
Chemical Name or Material Silica Standard, 1000ppm (1mL = 1mg SiO2)
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature