missing translation for 'onlineSavingsMsg'
Learn More

Fisher Chemical™ Silica Gel Sorbent (100-200 Mesh)

Catalog No. S8171
Change view
Click to view available options
Grade:
923
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S8171 923
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S8171 Supplier Fisher Chemical™ Supplier No. FLS8171
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

  • Grade 923
  • Pore Size: 32Å
  • Silica content: 99% Minimum
  • CAS: 7631-86-9

 

Specifications

Particle Size 75 to 150 μm
Grade 923
Packaging Amber Glass
Quantity 1 kg
Material Silica Gel