missing translation for 'onlineSavingsMsg'
Learn More

Silica Gel, Grade 47 (10-18 Mesh/Certified ACS), Fisher Chemical™

Catalog No. S161212
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S161212 2.5 kg Poly Bottle
S161500 500 g Poly Bottle
S16110 10 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S161212 Supplier Thermo Fisher Scientific Supplier No. S161212
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Silica Gel
Color Blue
Physical Form Solid
Name Note Tel-Tale™ Desiccant, Indicating, 10-18 Mesh
CAS 7646-79-9, 7631-86-9
Material Silica Gel
Quantity 2.5 kg
Identification Pass Test
Molecular Formula SiO2
MDL Number MFCD00011232
Mesh Size 10 to 18 Mesh
Packaging Poly Bottle
Suitability Pass Test
Grade Certified ACS
Show More Show Less