missing translation for 'onlineSavingsMsg'
Learn More

Silica gel desiccant, indicating, ACS

Catalog No. AA3673736
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3673736 500 g
AA3673722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3673736 Supplier Thermo Scientific Chemicals Supplier No. 03673736

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used to dry the air in industrial compressed air systems. It is also used as a preservation tool to control relative humidity in museum and library exhibitions and storage. Further, it is employed in diagnostic test strips, inhalation devices, syringes, drug test kits and hospital sanitation kits. | CAS: 7631-86-9 | O2Si | 60.08 g/mol

Silica gel desiccant is used to dry the air in industrial compressed air systems. It is also used as a preservation tool to control relative humidity in museum and library exhibitions and storage. Further, it is employed in diagnostic test strips, inhalation devices, syringes, drug test kits and hospital sanitation kits. In addition to this, it is used as a stationary phase in chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
PubChem CID 24261
ChEBI CHEBI:30563
SMILES O=[Si]=O

Specifications

Physical Form -6+18 Mesh Granules
Material Silica Gel
Quantity 500 g
Mesh Size −6+18 Mesh Granules
Packaging Plastic Bottle
Solubility Information Soluble in alkalies. Insoluble in water.
Sensitivity Hygroscopic
IUPAC Name dioxosilane
Grade ACS
Chemical Name or Material Silica gel desiccant
Hazard Category H350i-H360F
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 231-545-4
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only