missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Silica gel, for chromatography, 0.060-0.200 mm, 40 A
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For column chromatography
Specifications
Specifications
| Pore Size | 40 Å |
| Packaging | Plastic Drum |
| Quantity | 5 kg |
| Type | Silica Gel |
| Material | Silica Gel |
| Solubility Information | Solubility aq. soln.: insoluble. |
| Formula Weight | 60.08 |
| Chemical Name or Material | Silica gel |
RUO – Research Use Only