missing translation for 'onlineSavingsMsg'
Learn More

Silica gel, for chromatography, 0.075-0.250 mm, 150 A

Catalog No. AC419282500
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC419282500 250 g Glass bottle
AC419280010 1 kg Glass bottle
AC419280050 5 kg Plastic Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC419282500 Supplier Thermo Scientific Chemicals Supplier No. 419282500

CAS: 7631-86-9 | O2Si | 60.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For column chromatography

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
PubChem CID 24261
ChEBI CHEBI:30563
SMILES O=[Si]=O

Specifications

Color White
Physical Form Very Fine Granular Powder
Packaging Glass bottle
Particle Size Nominal particle size: >60 mesh: <7.5%, <200 mesh: <10.0%
Specific Surface Area 275 to 375 m2/g (BET)
Quantity 250 g
Merck Index 15, 8632
Solubility Information Solubility in water: insoluble.
IUPAC Name silanedione
Formula Weight 60.08
Chemical Name or Material Silica gel, For column chromatography
Show More Show Less

RUO – Research Use Only