Learn More
Thermo Scientific Chemicals Silica gel, C18-RP, 23%C, approx. 1 mmol/g, part. size: 40-63 micron
Catalog No. AC360112500
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC360112500 250 g
AC360110500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC360112500 Supplier Thermo Scientific Chemicals Supplier No. 360112500
Only null left
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For column chromatography
Specifications
Packaging Glass Bottle
Quantity 250 g
Material Silica Gel
What is the pore size of Silica gel, C18-RP, 23%C, ca.1.2 mmol/g, part. size 40-63 μm (Cat. No. 360110500, 360112500) in Angstroms?
The pore size of Silica gel, C18-RP, 23%C, ca.1.2 mmol/g, part. size 40-63 µm (Cat. No. 360110500, 360112500) is 60 Angstroms.
RUO – Research Use Only