missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Silibinin

Numéro de catalogue. AC463030250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AC463030250 25 g
AC463030010 1 g
AC463030050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. AC463030250 Fournisseur Thermo Scientific Chemicals Code fournisseur 463030250
Il en reste null

CAS: 22888-70-6 | C25H22O10 | 482.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 22888-70-6
Molecular Formula C25H22O10
Molecular Weight (g/mol) 482.44
InChI Key SEBFKMXJBCUCAI-UHFFFAOYNA-N
IUPAC Name 3,5,7-trihydroxy-2-[3-(4-hydroxy-3-methoxyphenyl)-2-(hydroxymethyl)-2,3-dihydro-1,4-benzodioxin-6-yl]-3,4-dihydro-2H-1-benzopyran-4-one
SMILES COC1=CC(=CC=C1O)C1OC2=CC(=CC=C2OC1CO)C1OC2=CC(O)=CC(O)=C2C(=O)C1O

Spécifications

Melting Point 164.0°C to 174.0°C
Quantity 25 g
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: soluble in acetone, ethyl acetate, methanol,, ethanol, sparingly soluble in chcl3
Formula Weight 482.44
Chemical Name or Material Silibinin
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

If eye irritation persists: Get medical advice/attention.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wear protective gloves/protective clothing/eye protection/face protection.

If skin irritation occurs: Get medical advice/attention.

EINECSNumber 245-302-5

RUO – Research Use Only