missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ SignalBoost™ Immunoreaction Enhancer Kit

Catalog No. 4072071kit
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4072071KIT 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4072071KIT Supplier MilliporeSigma™ Supplier No. 4072071KIT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed to enhance desired signal in assays such as immunoblotting, dot blotting, ELISA.

EMD Millipore Calbiochem™ SignalBoost™ Immunoreaction Enhancer Kit significantly improves sensitivity, protein targets of low immunoreactivity or low expression levels.

  • Replaces conventional antibody diluents
  • No additional assay time required
  • Minimizes background resulting in high signal to noise ratios
  • Used with nitrocellulose or PVDF membranes
  • Compatible with chemiluminescent or colorimetric detection systems
  • Convenient to use, no dilution needed
  • Packaging: Glass Bottle
  • Applications: Immunoassays, ELISA

Specifications

Description Immunoreaction Enhancer Kit
Quantity 1 Kit
Content And Storage Solution 1 for Primary Antibodies, Solution 2 for Secondary Antibodies, and user protocol. 2°C to 8°C