missing translation for 'onlineSavingsMsg'
Learn More

Sigma Pseudo™ Narcotic Scent (Heroin Formulation Powder), MilliporeSigma™ Supelco™

$125.43 - $989.43

Specifications

CAS 50-78-2
Product Line Sigma Pseudo
Physical Form Powder
MDL Number MFCD00165842
Type Standard
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111001677
View Documents
Merck Emd Millipore
P2548100G
100 g N/A
Only null left
 
111001676
View Documents
Merck Emd Millipore
P25481KG
1 kg N/A
Only null left
 
Description

Description

Specifications

Specifications

50-78-2
Powder
Standard
Sigma Pseudo
MFCD00165842
Videos
SDS
Documents

Documents

Sigma Pseudo is a trademark of Sigma-Aldrich Co. LLC