Learn More
ACROBiosystems Inc ACROBiosystems Mouse Siglec-10 Protein, His Tag
Numéro de catalogue. 501956547
Encompass
Recombinant Protein;1MG;Mouse Siglec-10, His Tag (SI0-M52H7) is expressed from human 293 cells (HEK293). It contains AA Met 18 - Lys 543 (Accession # Q80ZE3-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 60.6 kDa.
Numéro de catalogue. 501956547 Fournisseur ACROBiosystems Inc Code fournisseur SI0M52H71MG
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Recombinant Protein;1MG;Mouse Siglec-10, His Tag (SI0-M52H7) is expressed from human 293 cells (HEK293). It contains AA Met 18 - Lys 543 (Accession # Q80ZE3-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 60.6 kDa.