Learn More
Dräger™ Short-Term Detector Tubes - De Tube Hydrogen Sulfide
For gas detection
Supplier:  Dräger™ 8101461
 View more versions of this product
Catalog No. 19038824
Only null left
Description
Description
  • Glass vial
  • Calibrated 100mL sample of air drawn through tube with Drager Accuro™ bellows pump
Specifications
Specifications
Hydrogen Sulfide
0.2 to 5ppm
10/Box
Product Suggestions
Product Suggestions