missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Shikimic Acid, 98%

Catalog No. p-3737364
Change view
Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC132700010 1g Glass bottle
AC132700050 5g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC132700010 Supplier Thermo Scientific™ Supplier No. 132700010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 138-59-0
Molecular Formula C7H10O5
Molecular Weight (g/mol) 174.15
MDL Number MFCD00066278
InChI Key JXOHGGNKMLTUBP-HSUXUTPPSA-M
Synonym shikimic acid, shikimate, --shikimic acid, l-shikimic acid, 3r,4s,5r-3,4,5-trihydroxycyclohex-1-enecarboxylic acid, 3r,4s,5r-3,4,5-trihydroxycyclohex-1-ene-1-carboxylic acid, --shikimate, ccris 7681, unii-29ms2wi2nu, 3alpha,4alpha,5beta-trihydroxy-1-cyclohexene-1-carboxylic acid
PubChem CID 7057976
ChEBI CHEBI:36208
IUPAC Name (3R,4S,5R)-3,4,5-trihydroxycyclohexene-1-carboxylate
SMILES C1C(C(C(C=C1C(=O)[O-])O)O)O

Specifications

Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1g
Beilstein 10, 458
Merck Index 15, 8619
Specific Rotation Condition − 180.00 (25.00°C c=4,H2O)
Specific Rotation − 180.00
Solubility Information Solubility in water: 18g/100mL (20 c). Other solubilities: 2.25g/100mL abs.alcohol, 0.015g/100mL anhydr.ether, practically insoluble in chloroform, benzene and, petroleum ether
Formula Weight 174.15
Percent Purity 98%
Chemical Name or Material Shikimic acid
Show More Show Less
EINECSNumber 205-334-2

RUO – Research Use Only