missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Sheep anti-Bovine IgM Secondary Antibody, HRP

Catalog No. PA184688
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA184688 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA184688 Supplier Invitrogen™ Supplier No. PA184688
  • ONLINE EXCLUSIVE PRICE
    $820.49
    $477.00 / Each

    Save $343.49

    42.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sheep Polyclonal Secondary Antibody

Sheep anti bovine IgM recognizes bovine IgM and shows no cross-reactivity with other bovine immunoglobulin classes in immunoelectrophoresis. Sheep anti bovine IgM may cross-react with IgM from other species.

Anti-Bovine secondary antibodies are affinity-purified antibodies with well-characterized specificity for bovine (cow, ox) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Bovine IgM
Applications ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate HRP
Formulation PBS with 0.2% BSA and 0.05% ProClin 300
Host Species Sheep
Immunogen Purified bovine IgM.
Purification Method Affinity chromatography
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Bovine
Content And Storage 4°C, do not freeze
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less