Learn More
Epredia™ Shandon™ Plated Scalpel Blade Handle, Holds No. 20-25 Blades
Catalog No. 3810861
Change view
Click to view available options
Size:
# 4
Catalog No. Size
3810861 # 4
1 options
Catalog No. 3810861 Supplier Epredia™ Supplier No. SH5324
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Securely hold blades with the Epredia™ Shandon™ Plated Scalpel Blade Handle, covered in nickel-plated steel #4.
  • Properly hardened, tempered and passivated
  • Electronically polished and cleaned for a high-quality finish
  • Quality patterns, dimensions and finish held to exacting standards, producing the finest instruments available
  • Defect-free in material and workmanship
Specifications
Blade Material Nickel-plated Steel
Size # 4
For Use With (Equipment) # 20 to # 25 Blade for Deep Surgery