Learn More
Epredia™ Cytospin™ Collection Fluid
Catalog No. 6768315
Change view
Click to view available options
Quantity:
120mL
500mL
4 L
10L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6768315 500mL
9990323 120mL
6768001 4 L
2860179 10L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 6768315 Supplier Epredia™ Supplier No. 6768315
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Store specimens or unstained slides for weeks without adverse effect with Epredia™ Shandon™ Cytospin™ Collection Fluid.
  • Packaged in convenient pump bottle or pre-filled specimen cups
  • Ready-to-use alcohol/Carbowax™ (PEG) combination for direct specimen collection and fixation
Specifications
Product Type Collection Fluid
Quantity 500mL
Solution Type Alcohol/Carbowax (PEG) combination