missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ SGE Autosampler Syringes, Cemented Needle, 10μL
For use with CTC/LEAP Autosamplers
Supplier: MilliporeSigma™ Supelco™ 29626U
Specifications
| Stainless Steel (Tip) | |
| 0.47 mm | |
| 1 | |
| 10 μL | |
| Cone | |
| Cemented |
| 26 G | |
| Syringe | |
| 6 Pk. | |
| 50 mm | |
| CTC, LEAP |