missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Seward™ Stomacher™ Model 80 Biomaster Blender 110V
Description
Compact size, reliability and performance has made the Seward Stomacher 80 Biomaster a standard processing tool in bioscience, healthcare and veterinary, processing a range of samples from plant and animal tissue to china clay; Process samples of bio hazardous nature as sample is contained at all times in sterile plastic bag; Prevents aerosol release into the lab and cross contamination of samples; Small size allows it to fit conveniently into safety cabinets; sample capacity 5-80ml; adjustable processing time and paddle speed; 5000 fold improvement in cell recovery and viability from traditional methods
Specifications
Specifications
| Speed | 230, 265, 300 rpm |
| Voltage | 110 V |
| Electrical Requirements | 110 V, 65 Hz |
| Wattage | 75 W |
| Volume Range (English) | 0.16 to 2.7 oz. |
| Volume Range (Metric) | 5 to 80 mL |
| Depth (English) | 12.7 in. |
| Depth (Metric) | 32.5 cm |
| Dimensions (L x W x H) | 32.5 x 29 x 19 cm |
| Height (English) | 7.5 in. |
| Show More |